Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)/Cy5®-PE (Phycoerythrin) [clone: 5F4/5F22/5F41]
Katalog # USBIC2254-87
Leverantör: US Biological
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:CD3
- Klonalitet:Monoclonal
- Klon:5F4/5F22/5F41
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Flödescytometri:Yes
- Värd:Mouse
- Isotyp:IgG1
- Reaktivitet:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide. Triple color combination consisting of FITC
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:25 Tests
- Förp.:25 Tests
Specifikationer
Om denna produkt
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 5F4/5F22/5F41
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)/Cy5®-PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1+IgG1+IgG1
Reactivity: Human