Order Entry
Sweden
ContactUsLinkComponent
 
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)
  ROCK813-4302
undefined
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)
  ROCK813-4302

 

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgG (H+L)
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Peroxidase
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Sheep
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • Mått:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Sheep IgG whole molecule
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    500 µg
  • Förp.:
    500 µG

 

 

Suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:4000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Sheep