Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Katalog # ROCK813-4002
undefined
Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Katalog # ROCK813-4002

Specifikationer

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgG (H+L)
  • Klonalitet:
    Polyclonal
  • Konjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • Värd:
    Rabbit
  • Reaktivitet:
    Sheep
  • Korsad adsorption:
    none
  • Mått:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Sheep IgG whole molecule
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    500 µl
  • Förp.:
    500 µG

Specifikationer

Om denna produkt

Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Sheep