Order Entry
Sweden
ContactUsLinkComponent
Anti-IgM µ Rabbit Polyclonal Antibody (Texas Red®)
Anti-IgM µ Rabbit Polyclonal Antibody (Texas Red®)
  ROCK612-4907
undefined
Anti-IgM µ Rabbit Polyclonal Antibody (Texas Red®)
  ROCK612-4907

 

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgM µ
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Texas Red®
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Gelskifte:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Immunutfällning:
    Yes
  • Reaktivitet:
    Rat
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Mått:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Rat IgM mu heavy chain
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    1,5 mg
  • Förp.:
    1.500 µG

 

 

Secondary Rabbit Anti-IgM (mu chain) Reacts with Rat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: µ-chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat