Order Entry
Sweden
ContactUsLinkComponent
Anti-IgM µ Goat Polyclonal Antibody (Biotin)
Anti-IgM µ Goat Polyclonal Antibody (Biotin)
Catalog # ROCK612-1607
undefined
Anti-IgM µ Goat Polyclonal Antibody (Biotin)
Catalog # ROCK612-1607

Specifikationer

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgM µ
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Biotin
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Gelskifte:
    Yes
  • Värd:
    Goat
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Immunutfällning:
    Yes
  • Reaktivitet:
    Rat
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Mått:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Rat IgM mu heavy chain
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    1 mg
  • Förp.:
    1 mg

Specifikationer

Om denna produkt

This product has been assayed against 1.0 µg of Rat IgM in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room tempertaure.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Rat