Order Entry
Sweden
ContactUsLinkComponent
Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # ROCK612-1507
undefined
Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # ROCK612-1507

Specifikationer

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgM µ
  • Klonalitet:
    Polyclonal
  • Konjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Gelskifte:
    Yes
  • Värd:
    Goat
  • ImmunKemi:
    Yes
  • Immunutfällning:
    Yes
  • Reaktivitet:
    Rat
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Mått:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Rat IgM mu heavy chain
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    1 mg
  • Förp.:
    1 mg

Specifikationer

Om denna produkt

This product has been assayed against 1.0 µg of Rat IgM in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature.  A working dilution of 1:500 to 1:2,000 is suggested for this product.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Rat