Anti-IgG (H+L) Guniea pig Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalog # ROCK611-202-122
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Sekundär
- Antigensymbol:IgG (H+L)
- Klonalitet:Polyclonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Värd:Guinea pig
- ImmunoFluorescens:Yes
- Reaktivitet:Rabbit
- Korsad adsorption:Human, Goat, Mouse
- Mått:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free
- Förvaringstemperatur:Cool
- Koncentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Rabbit IgG whole molecule
- Rening:purified by immunoaffinity chromatography
- Storlek:1 mg
- Förp.:1 mg
Specifikationer
Om denna produkt
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Guinea Pig
Isotype:
Reactivity: Rabbit