Order Entry
Sweden
ContactUsLinkComponent
 
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # ROCK611-1502
undefined
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # ROCK611-1502

Specifikationer

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgG (H+L)
  • Klonalitet:
    Polyclonal
  • Konjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Gelskifte:
    Yes
  • Värd:
    Goat
  • ImmunKemi:
    Yes
  • Immunutfällning:
    Yes
  • Reaktivitet:
    Rabbit
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Mått:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-Rabbit IgG (H&L) was produced by repeated immunization with rabbit whole IgG molecule in goat.
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    1 mg
  • Förp.:
    1 mg

Specifikationer

Om denna produkt

Anti-Rabbit IgG alkaline phosphatase conjugated antibody is suitable for immunoblotting (western or dot blot), ELISA, immunochemistry as well as other alkaline phosphatase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:30,000; Western Blot: 1:1000 - 1:4000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope: Heavy and Light chain
Host:
Isotype:
Reactivity: