Anti-IgG (H+L) Chicken Polyclonal Antibody (Texas Red®)
Katalog # ROCK605-9902
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Sekundär
- Antigensymbol:IgG (H+L)
- Klonalitet:Polyclonal
- Konjugation:Texas Red®
- Värd:Chicken
- ImmunoFluorescens:Yes
- Reaktivitet:Goat
- Korsad adsorption:none
- Mått:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Förvaringstemperatur:Cool
- Koncentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Goat IgG whole molecule
- Rening:purified by immunoaffinity chromatography
- Storlek:1 mg
- Förp.:1 mg
Specifikationer
Om denna produkt
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Chicken
Isotype:
Reactivity: Goat