Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG F(c) Goat Polyclonal Antibody
Anti-IgG F(c) Goat Polyclonal Antibody
Katalog # ROCK604-1103
undefined
Anti-IgG F(c) Goat Polyclonal Antibody
Katalog # ROCK604-1103

Specifikationer

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgG F(c)
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Gelskifte:
    Yes
  • Värd:
    Goat
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Immunutfällning:
    Yes
  • Reaktivitet:
    Dog
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Mått:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    2,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Dog IgG F(c) fragment
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    2 mg
  • Förp.:
    2 mg

Specifikationer

Om denna produkt

Secondary Goat Anti-IgG F(c) Reacts with Dog (Canine)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Dog