Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # ROCK603-105-007
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Sekundär
- Antigensymbol:IgM µ
- Klonalitet:Polyclonal
- Konjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Värd:Goat
- ImmunKemi:Yes
- Reaktivitet:Chicken
- Western blot:Yes
- Korsad adsorption:none
- Mått:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
- Förvaringstemperatur:Cool
- Koncentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Chicken IgM mu heavy chain
- Rening:purified by immunoaffinity chromatography
- Storlek:1 mg
- Förp.:1 mg
Specifikationer
Om denna produkt
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1:200 - 1:2000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Chicken