Order Entry
Sweden
ContactUsLinkComponent
Anti-KLH Rabbit Polyclonal Antibody (Biotin)
Anti-KLH Rabbit Polyclonal Antibody (Biotin)
Catalog # ROCK600-406-466
undefined
Anti-KLH Rabbit Polyclonal Antibody (Biotin)
Catalog # ROCK600-406-466

Specifications

  • Antikroppstyp:
    Märke
  • Antikroppsnamn:
    Keyhole Limpet Hemocyanin
  • Antigensymbol:
    KLH
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Biotin
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Goat
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • Mått:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    KLH (Keyhole Limpet Hemocyanin
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifications

About this item

Anti-KLH Antibody is suitable for western blotting to detect a single band of the expected apparent molecular weight and for ELISA. Anti-KLH antibody biotin conjugate is suitable for use with streptavidin reporter systems.

Recommended Dilutions: ELISA: 1:250000; Western Blot: 1:5000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: