Order Entry
Sweden
ContactUsLinkComponent
Anti-TF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-TF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # ROCK600-403-034
undefined
Anti-TF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # ROCK600-403-034

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    TRANSFERRIN
  • Klonalitet:
    Polyclonal
  • Konjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • Mått:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Human Transferrin
  • Rening:
    purified by Protein A Sepharose Fast Flow followed by POROS EP antigen resin affinity purification
  • Storlek:
    500 µg
  • Förp.:
    500 µG

Specifikationer

Om denna produkt

Recommended Dilutions: ELISA: 1:3000 - 1:12000; Western Blot: 1:300 - 1:1000;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Primary
Antigen: TF
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human