Order Entry
Sweden
ContactUsLinkComponent
Anti-GST Goat Polyclonal Antibody (Texas Red®)
Anti-GST Goat Polyclonal Antibody (Texas Red®)
  ROCK600-109-200
undefined
Anti-GST Goat Polyclonal Antibody (Texas Red®)
  ROCK600-109-200

 

  • Antikroppstyp:
    Märke
  • Antikroppsnamn:
    Glutathione-S-Transferase
  • Antigensymbol:
    GST
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Texas Red®
  • ELISA:
    Yes
  • Värd:
    Goat
  • ImmunoFluorescens:
    Yes
  • Reaktivitet:
    Cat
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Storlek:
    1 mg
  • Förp.:
    1 mg

 

 

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype:
Reactivity: