Anti-A2M Goat Polyclonal Antibody
Katalog # ROCK200-101-207S
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Alpha-2-macroglobulin
- Antigensymbol:A2M
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Goat
- ImmunKemi:Yes
- Reaktivitet:Human
- Western blot:Yes
- ChIP:Yes
- Antigen synonymer:CPAMD5
- Storlek:25 µl
- Förp.:25 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: A2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human