Anti-Dog serum Rabbit Polyclonal Antibody
Catalog # ROCK104-4101
Supplier: Rockland Immunochemicals
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Dog Serum
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Immunutfällning:Yes
- Reaktivitet:Dog
- Western blot:Yes
- Korsad adsorption:none
- Mått:Antiserum
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Förvaringstemperatur:Cool
- Koncentration:90 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Dog serum Proteins
- Rening:delipidation and defibrination
- Storlek:2 ml
- Förp.:2 mL
Specifications
About this item
Anti-Canine serum is suitable for the specific deteciton of protiens found in dog serum. Cross reactivity to serum-protiens from other species has not been tested. Optimum experimental usage should be determined by researcher.
Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: Dog serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog