Anti-RFX5 Rabbit Polyclonal Antibody
Katalog # ROCK100-401-193
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Regulatory factor X, 5 (influences HLA class II expression)
- Antigensymbol:RFX5
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Immunutfällning:Yes
- Reaktivitet:Human
- Western blot:Yes
- Epitop:near the N-terminus
- Korsad adsorption:none
- Mått:Antiserum
- Form:Liquid (sterile filtered)
- Gen-ID:5993
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Förvaringstemperatur:Cool
- Koncentration:90 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:RFX5 (N-terminal specific) peptide corresponding to a region near the N-terminus of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Rening:delipidation and defibrination
- Storlek:100 µl
- Förp.:100 µl
Specifikationer
Om denna produkt
This product was also tested in a gel supershift assay and found to be reactive against RFX5 complexes using 0.5 to 1.0 µl per assay. This product was also tested in a gel supershift assay and found to be reactive against RFX5 complexes.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: RFX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human