Order Entry
Sweden
ContactUsLinkComponent
 
Anti-GLPK Goat Polyclonal Antibody
  ROCK100-101-211
undefined
Anti-GLPK Goat Polyclonal Antibody
  ROCK100-101-211

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    glycerol kinase (Cellulomonas species)
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Goat
  • Immunutfällning:
    Yes
  • Reaktivitet:
    Cellulomonas
  • Western blot:
    Yes
  • Korsad adsorption:
    none
  • Mått:
    Antiserum
  • Form:
    Lyophilized
  • Gen-ID:
    6960048
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Förvaringstemperatur:
    Cool
  • Koncentration:
    90 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    Glycerol Kinase (Cellulomonas species)
  • Rening:
    delipidation and defibrination
  • Storlek:
    2 ml
  • Förp.:
    2 mL

 

 

This product has been assayed against 1.0 µg of Glycerol Kinase (Celullomonas species) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: GLPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Cellulomonas