Anti-Cyr61/CCN1 Rabbit Polyclonal Antibody (DyLight 488)
Catalog # NOVUNB100-356G
Supplier: Novus Biologicals
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Cysteine-rich, Angiogenic Inducer 61
- Antigensymbol:CYR61
- Klonalitet:Polyclonal
- Konjugation:DyLight® 488
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Reaktivitet:Human
- Western blot:Yes
- Gen-ID:3491
- Antigen synonymer:CCN1|IGFBP10|GIG1
- Storage buffer:50 mM Sodium Borate
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:100 µl
- Förp.:0,1 mL
Specifications
About this item
The Cyr61/CCN1 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to Cyr61/CCN1. This antibody reacts with human, rabbit. The Cyr61/CCN1 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: Cyr61/CCN1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit