Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Friend leukemia virus integration 1
- Antigensymbol:FLI1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Epitop:N-Terminal
- Korsad adsorption:No
- Form:Lyophilized
- Antigen synonymer:Friend leukemia virus integration 1|transcription factor ERGB|Fli-1|FLI1|ERGB transcription factor|Fli-1 proto-oncogene
- Aminosyretal:54 to 73
- UniProtKB:Q01543
- Sequence:QQEWINQPVRVNVKREYDHM
- Förvaringstemperatur:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human FLI1 (54-73aa QQEWINQPVRVNVKREYDHM), identical to the related rat and mouse sequences.
- Rening:purified by immunogen affinity purification
- Storlek:100 µg
- Förp.:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Friend leukemia integration 1 transcription factor(FLI1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Friend leukemia integration 1 transcription factor, also known as Transcription factor ERGB or Proto-oncogene Fli-1 is a protein that in humans is encoded by the human FLI1 gene. The fli1 protooncogene encodes a member of the ETS family of winged helix-turn-helix transcription factors. This gene is mapped to 11q24.3. Fli-1 is activated through retroviral insertional mutagenesis in 90% of F-MuLV-induced erythroleukemias. The constitutive activation of fli-1 in erythroblasts leads to a dramatic shift in the Epo/Epo-R signal transduction pathway, blocking erythroid differentiation, activating the Ras pathway, and resulting in massive Epo-independent proliferation of erythroblasts. These results suggest that Fli-1 overexpression in erythroblasts alters their responsiveness to Epo and triggers abnormal proliferation by switching the signaling event(s) associated with terminal differentiation to proliferation.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: FLI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat