Order Entry
Sweden
ContactUsLinkComponent
Anti-EIF4EBP1 Rabbit polyclonal antibody unconjugated
Anti-EIF4EBP1 Rabbit polyclonal antibody unconjugated
Catalog # ABCAAB47365-100
Supplier:  Abcam
Anti-EIF4EBP1 Rabbit polyclonal antibody unconjugated
Catalog # ABCAAB47365-100
Supplier:  Abcam

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Eukaryotic translation initiation factor 4E binding protein 1
  • Antigensymbol:
    EIF4EBP1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human