Order Entry
Sweden
ContactUsLinkComponent
Anti-IL5 Mouse Monoclonal Antibody [clone: QS-5]
Anti-IL5 Mouse Monoclonal Antibody [clone: QS-5]
Catalog # ABCAAB25034-100
Leverantör:  Abcam
Anti-IL5 Mouse Monoclonal Antibody [clone: QS-5]
Catalog # ABCAAB25034-100
Leverantör:  Abcam

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Interleukin 5
  • Antigensymbol:
    IL5
  • Klonalitet:
    Monoclonal
  • Klon:
    QS-5
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG1
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Neutralisation:
    Yes
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL5 (interleukin 5 (colony-stimulating factor, eosinophil))
Clonality: monoclonal
Clone: QS-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human