Order Entry
Sweden
ContactUsLinkComponent
Anti-NCOA1 Rabbit Polyclonal Antibody
Anti-NCOA1 Rabbit Polyclonal Antibody
  ABCAAB10308-50
 :  Abcam
Anti-NCOA1 Rabbit Polyclonal Antibody
  ABCAAB10308-50
 :  Abcam

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Nuclear receptor coactivator 1
  • Antigensymbol:
    NCOA1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Immunutfällning:
    Yes
  • Reaktivitet:
    Human,
    Mouse
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Storlek:
    50 µg
  • Förp.:
    50 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NCOA1 (nuclear receptor coactivator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse