Order Entry
Sweden
ContactUsLinkComponent
Anti-TNF Rabbit Polyclonal Antibody
Anti-TNF Rabbit Polyclonal Antibody
  ABCAAB9739-100
 :  Abcam
Anti-TNF Rabbit Polyclonal Antibody
  ABCAAB9739-100
 :  Abcam

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    tumor necrosis factor
  • Antigensymbol:
    TNF
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Neutralisation:
    Yes
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Storlek:
    100 µg
  • Förp.:
    100 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse