Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # ABCAAB7097-500
Leverantör: Abcam
Specifikationer
- Antikroppstyp:Sekundär
- Antigensymbol:IgG
- Klonalitet:Polyclonal
- Konjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Värd:Goat
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Western blot:Yes
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:500 µg
- Förp.:500 µG
Specifikationer
Om denna produkt
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: