Anti-IgG Donkey Polyclonal Antibody (Cy3.5®)
Katalog # ABCAAB6950-500
Leverantör: Abcam
Specifikationer
- Antikroppstyp:Sekundär
- Antigensymbol:IgG
- Klonalitet:Polyclonal
- Konjugation:Cy3.5®
- ELISA:Yes
- Flödescytometri:Yes
- Värd:Donkey
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:500 µg
- Förp.:500 µG
Specifikationer
Om denna produkt
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy3.5®
Epitope:
Host: Donkey
Isotype: none
Reactivity: