Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ABCAAB6915-1
Supplier:  Abcam
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ABCAAB6915-1
Supplier:  Abcam

Specifications

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgG
  • Klonalitet:
    Polyclonal
  • Konjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Värd:
    Goat
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Western blot:
    Yes
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Storlek:
    1 mg
  • Förp.:
    1 mg

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: