Order Entry
Sweden
ContactUsLinkComponent
Anti-CALM1 Mouse Monoclonal Antibody [clone: 2D1]
Anti-CALM1 Mouse Monoclonal Antibody [clone: 2D1]
Katalog # ABCAAB2860-200
Leverantör:  Abcam
Anti-CALM1 Mouse Monoclonal Antibody [clone: 2D1]
Katalog # ABCAAB2860-200
Leverantör:  Abcam

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    calmodulin 1 (phosphorylase kinase, delta)
  • Antigensymbol:
    CALM1
  • Klonalitet:
    Monoclonal
  • Klon:
    2D1
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Värd:
    Mouse
  • Isotyp:
    IgG1
  • Western blot:
    Yes
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Storlek:
    200 µl
  • Förp.:
    200 µl

Specifikationer

Om denna produkt

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CALM1 (calmodulin 1 (phosphorylase kinase, delta))
Clonality: monoclonal
Clone: 2D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: