Anti-Adducin Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: Adducin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- BIRBORB10044-100
- BIRBORB10044-200
- Antigensymbol:
- ADD
- ADD
- Antikroppsnamn:
- Adducin
- Adducin
- Antikroppstyp:
- Primär
- Primär
- Klonalitet:
- Polyclonal
- Polyclonal
- Konjugation:
- Unconjugated
- Unconjugated
- Värd:
- Rabbit
- Rabbit
- Isotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunKemi:
- Yes
- Yes
- Immunogen:
- Synthetic peptide
- Synthetic peptide
- Form:
- liquid
- liquid
- Molekylvikt:
- 180 kDa
- 180 kDa
- Rening:
- purified by affinity chromatography
- purified by affinity chromatography
- Koncentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Förvaringstemperatur:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice