Anti-KLF2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Katalog # BIRBORB9120-100
Leverantör: Biorbyt
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Kruppel-like factor 2
- Antigensymbol:KLF2
- Klonalitet:Polyclonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molekylvikt:37 kDa
- Förvaringstemperatur:Cool
- Koncentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human KLF2.
- Rening:purified by affinity chromatography
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: KLF2 (Kruppel-like factor 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: