Order Entry
Sweden
ContactUsLinkComponent
 
Anti-GSTT1 Rabbit Polyclonal Antibody
  ORIGTA313305
 :  OriGene
Anti-GSTT1 Rabbit Polyclonal Antibody
  ORIGTA313305
 :  OriGene

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Glutathione S-transferase theta 1
  • Antigensymbol:
    GSTT1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    internal
  • Korsad adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Rening:
    purified by affinity chromatography
  • Förp.:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: GSTT1 (glutathione S-transferase theta 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human