Anti-RUNX1 Rabbit Polyclonal Antibody
ORIGTA313288
: OriGene
- Antikroppstyp:Primär
- Antikroppsnamn:Runt-related Transcription Factor 1
- Antigensymbol:RUNX1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Korsad adsorption:none
- Form:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Rening:purified by affinity chromatography
- Förp.:100 µl
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000
Type: Primary
Antigen: RUNX1 (runt-related transcription factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat