Anti-SMOX Rabbit Polyclonal Antibody
AVIVARP65968_P050
- Antikroppstyp:Primär
- Antikroppsnamn:spermine oxidase
- Antigensymbol:SMOX
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:54498
- Antigen synonymer:PAO-1|PAO|C20orf16|PAOH1|PAOH|SMO|PAO1
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: SMOX (spermine oxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human