Anti-NDRG4 Rabbit Polyclonal Antibody
AVIVARP64737_P050
- Antikroppstyp:Primär
- Antikroppsnamn:NDRG family member 4
- Antigensymbol:NDRG4
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:65009
- Antigen synonymer:SMAP8|BDM1|SMAP-8
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: NDRG4 (NDRG family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: