Anti-IKBKE Rabbit Polyclonal Antibody
Catalog # AVIVARP63182_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Inhibitor Of Kappa Light Polypeptide Gene Enhancer in B-cells, kinase epsilon
- Antigensymbol:IKBKE
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:9641
- Antigen synonymer:IKKE|IKKI|IKK-i|IKK-E
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: IKBKE (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: