Anti-MAPRE1 Rabbit Polyclonal Antibody
Katalog # AVIVARP61761_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:microtubule-associated protein, RP/EB family, member 1
- Antigensymbol:MAPRE1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:13589
- Antigen synonymer:AI462499|AI504412|AW260097|5530600P05Rik|Eb1|BIM1p|D2Ertd459e
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: MAPRE1 (microtubule-associated protein, RP/EB family, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: