Anti-XRCC3 Rabbit Polyclonal Antibody
Katalog # AVIVARP61275_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:X-ray repair complementing defective repair in Chinese hamster cells 3
- Antigensymbol:XRCC3
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:7517
- Antigen synonymer:CMM6
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: XRCC3 (X-ray repair complementing defective repair in Chinese hamster cells 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: