Anti-KNDC1 Rabbit Polyclonal Antibody
Katalog # AVIVARP60888_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:kinase non-catalytic C-lobe domain (KIND) containing 1
- Antigensymbol:KNDC1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:85442
- Antigen synonymer:bB439H18.3|C10orf23|RASGEF2
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: KNDC1 (kinase non-catalytic C-lobe domain (KIND) containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human