Anti-HNMT Rabbit Polyclonal Antibody
Katalog # AVIVARP59621_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Histamine N-methyltransferase
- Antigensymbol:HNMT
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:3176
- Antigen synonymer:HNMT-S1|HMT|HNMT-S2
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: HNMT (histamine N-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: