Anti-ALDH5A1 Rabbit Polyclonal Antibody
AVIVARP59587_P050
- Antikroppstyp:Primär
- Antikroppsnamn:aldehyde dehydrogenase 5 family, member A1
- Antigensymbol:ALDH5A1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:7915
- Antigen synonymer:SSDH|SSADH
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: ALDH5A1 (aldehyde dehydrogenase 5 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: