Anti-NAP1L4 Rabbit Polyclonal Antibody
Catalog # AVIVARP58308_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:nucleosome assembly protein 1-like 4
- Antigensymbol:NAP1L4
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:17955
- Antigen synonymer:2810410H14Rik|D7Wsu30e|AI316776|Nap2
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: NAP1L4 (nucleosome assembly protein 1-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: