Anti-PHLDA2 Rabbit Polyclonal Antibody
AVIVARP58237_P050
- Antikroppstyp:Primär
- Antikroppsnamn:Pleckstrin homology-like domain A2
- Antigensymbol:PHLDA2
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:7262
- Antigen synonymer:TSSC3|HLDA2|BRW1C|IPL|BWR1C
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: PHLDA2 (pleckstrin homology-like domain, family A, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human