Order Entry
Sweden
ContactUsLinkComponent
Anti-IMPA2 Rabbit Polyclonal Antibody
Anti-IMPA2 Rabbit Polyclonal Antibody
Katalog # AVIVARP58230_P050
Leverantör:  Aviva Systems Biology
undefined
Anti-IMPA2 Rabbit Polyclonal Antibody
Katalog # AVIVARP58230_P050
Leverantör:  Aviva Systems Biology

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    inositol (myo)-1(or 4)-monophosphatase 2
  • Antigensymbol:
    IMPA2
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Korsad adsorption:
    none
  • Gen-ID:
    114663
  • Antigen synonymer:
    2210415D20Rik|AW259601|AI326924
  • Förvaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Förp.:
    50 µG

Specifikationer

Om denna produkt

Type: Primary
Antigen: IMPA2 (inositol(myo)-1(or 4)-monophosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: