Anti-RAP1A Rabbit Polyclonal Antibody
Katalog # AVIVARP56195_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:RAP1A, Member Of RAS Oncogene Family
- Antigensymbol:RAP1A
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:109905
- Antigen synonymer:AI848598|Rap1|G-22K|Krev-1
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: RAP1A (RAP1A, member of RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: