Anti-NRROS Rabbit Polyclonal Antibody
Catalog # AVIVARP55907_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Negative regulator of reactive oxygen species
- Antigensymbol:NRROS
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:375387
- Antigen synonymer:ELLP3030|GARPL1|UNQ3030|LRRC33
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: NRROS (negative regulator of reactive oxygen species)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: