Anti-TRAPPC2 Rabbit Polyclonal Antibody
Katalog # AVIVARP50567_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Trafficking Protein Particle Complex 2
- Antigensymbol:TRAPPC2
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human,Rat
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:501550
- Antigen synonymer:RGD1306925
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: TRAPPC2 (trafficking protein particle complex 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat