Anti-SLC17A5 Rabbit Polyclonal Antibody
Catalog # AVIVARP43932_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Solute carrier family 17 (acidic sugar transporter), member 5
- Antigensymbol:SLC17A5
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:26503
- Antigen synonymer:AST, ISSD, NSD, SD, SIALIN, SIASD, SLD
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC17A5 (solute carrier family 17 (acidic sugar transporter), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: