Anti-SLC20A1 Rabbit Polyclonal Antibody
Katalog # AVIVARP43905_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:solute carrier family 20 (phosphate transporter), member 1
- Antigensymbol:Solute carrier family 20, member 1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:6574
- Antigen synonymer:GLVR1|Glvr-1|PiT-1|PIT1
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: SLC20A1 (solute carrier family 20 (phosphate transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: