Anti-RNF125 Rabbit Polyclonal Antibody
Katalog # AVIVARP43234_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Ring Finger Protein 125
- Antigensymbol:RNF125
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:54941
- Antigen synonymer:TRAC-1|TNORS|TRAC1
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: RNF125 (ring finger protein 125, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: