Anti-LTN1 Rabbit Polyclonal Antibody
Katalog # AVIVARP43201_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Listerin E3 ubiquitin protein ligase 1
- Antigensymbol:LTN1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:26046
- Antigen synonymer:RNF160|ZNF294|C21orf98|C21orf10
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: LTN1 (listerin E3 ubiquitin protein ligase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human